missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trivinylboroxin - Pyridine Complex 98.0+%, TCI America™

Supplier:  TCI America T24981G

 View more versions of this product

Catalog No. T24981G



Description

Description

Specifications

Specifications

2,4,6-Trivinylboroxin - Pyridine Complex
51°C
C11H14B3NO3
1 g
YLHJACXHRQQNQR-UHFFFAOYSA-N
pyridine;2,4,6-tris(ethenyl)-1,3,5,2,4,6-trioxatriborinane
2734806
≥98.0% (T)
95010-17-6
White-Yellow
MFCD03839440
2,4,6-trivinyl-1,3,5,2,4,6-trioxatriborinane compound with pyridine 1:1, trivinylboroxin pyridine complex, vinylboronic anhydride pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine, 2,4,6-trivinylcyclotriboroxane pyridine, vinylboronic anhydride pyridine, 2,4,6-trivinylcyclotriboroxane pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine complex, boroxin, triethenyl-, compd. with pyridine 1:1, pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
B1(OB(OB(O1)C=C)C=C)C=C.C1=CC=NC=C1
240.667
240.67
Crystalline Powder

Chemical Identifiers

95010-17-6
240.667
YLHJACXHRQQNQR-UHFFFAOYSA-N
2734806
B1(OB(OB(O1)C=C)C=C)C=C.C1=CC=NC=C1
C11H14B3NO3
MFCD03839440
2,4,6-trivinyl-1,3,5,2,4,6-trioxatriborinane compound with pyridine 1:1, trivinylboroxin pyridine complex, vinylboronic anhydride pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine, 2,4,6-trivinylcyclotriboroxane pyridine, vinylboronic anhydride pyridine, 2,4,6-trivinylcyclotriboroxane pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine complex, boroxin, triethenyl-, compd. with pyridine 1:1, pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
pyridine;2,4,6-tris(ethenyl)-1,3,5,2,4,6-trioxatriborinane
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions