missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(4-Chlorobenzoyl)pyridine 97.0+%, TCI America™
Supplier: TCI America C264725G
Specifications
2-(4-Chlorobenzoyl)pyridine | |
64°C | |
160°C | |
MFCD02930888 | |
4-Chlorophenyl 2-Pyridyl Ketone | |
ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1 | |
217.65 | |
217.65 | |
Crystalline Powder |
6318-51-0 | |
White-Yellow | |
C12H8ClNO | |
25 g | |
KHXSJSBQIWAIEG-UHFFFAOYSA-N | |
2-(4-chlorobenzoyl)pyridine | |
80594 | |
≥97.0% (GC,T) |
Chemical Identifiers
6318-51-0 | |
217.65 | |
KHXSJSBQIWAIEG-UHFFFAOYSA-N | |
80594 | |
ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1 |
C12H8ClNO | |
MFCD02930888 | |
4-Chlorophenyl 2-Pyridyl Ketone | |
2-(4-chlorobenzoyl)pyridine |
Safety and Handling
TSCA : No