missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Chlorobenzoyl)pyridine 97.0+%, TCI America™

$77.03 - $391.05

Chemical Identifiers

CAS 6318-51-0
Molecular Formula C12H8ClNO
Molecular Weight (g/mol) 217.65
MDL Number MFCD02930888
InChI Key KHXSJSBQIWAIEG-UHFFFAOYSA-N
Synonym 4-Chlorophenyl 2-Pyridyl Ketone
PubChem CID 80594
IUPAC Name 2-(4-chlorobenzoyl)pyridine
SMILES ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C26475G
View Documents
TCI America
C26475G
5 g
Each for $77.03
 
C264725G
View Documents
TCI America
C264725G
25 g
Each for $391.05
 
Description

Description

Specifications

Chemical Identifiers

6318-51-0
217.65
KHXSJSBQIWAIEG-UHFFFAOYSA-N
80594
ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1
C12H8ClNO
MFCD02930888
4-Chlorophenyl 2-Pyridyl Ketone
2-(4-chlorobenzoyl)pyridine

Specifications

6318-51-0
White-Yellow
C12H8ClNO
5 g
KHXSJSBQIWAIEG-UHFFFAOYSA-N
2-(4-chlorobenzoyl)pyridine
80594
≥97.0% (GC,T)
2-(4-Chlorobenzoyl)pyridine
64°C
160°C
MFCD02930888
4-Chlorophenyl 2-Pyridyl Ketone
ClC1=CC=C(C=C1)C(=O)C1=CC=CC=N1
217.65
217.65
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'