Learn More
2,4-Dibromobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1988703
Description
2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications
2,4-Dibromobenzenesulfonyl chloride | |
82°C to 86°C | |
C6H3Br2ClO2S | |
1 g | |
2694058 | |
2,4-dibromobenzene-1-sulfonyl chloride, 2,4-dibromobenzenesulphonyl chloride, 2,4-dibromophenyl chlorosulfone, pubchem5111, 2,4-dibromophenylsulfonyl chloride, benzenesulfonylchloride, 2,4-dibromo, 2,4-bis bromanyl benzenesulfonyl chloride, 2,4-dibromobenzenesulfonyl chloride | |
OMOXIKAWVFDFNX-UHFFFAOYSA-N | |
2,4-dibromobenzenesulfonyl chloride | |
2737366 | |
98% |
72256-95-2 | |
128°C (0.2 mmHg) | |
MFCD03093759 | |
UN3261 | |
Moisture sensitive | |
Insoluble in water. | |
ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1 | |
334.41 | |
334.41 |
Chemical Identifiers
72256-95-2 | |
334.41 | |
OMOXIKAWVFDFNX-UHFFFAOYSA-N | |
2737366 | |
ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1 |
C6H3Br2ClO2S | |
MFCD03093759 | |
2,4-dibromobenzene-1-sulfonyl chloride, 2,4-dibromobenzenesulphonyl chloride, 2,4-dibromophenyl chlorosulfone, pubchem5111, 2,4-dibromophenylsulfonyl chloride, benzenesulfonylchloride, 2,4-dibromo, 2,4-bis bromanyl benzenesulfonyl chloride, 2,4-dibromobenzenesulfonyl chloride | |
2,4-dibromobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only