Learn More
2,4-Dibromobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
$94.71 - $321.33
Chemical Identifiers
CAS | 72256-95-2 |
---|---|
Molecular Formula | C6H3Br2ClO2S |
Molecular Weight (g/mol) | 334.41 |
MDL Number | MFCD03093759 |
InChI Key | OMOXIKAWVFDFNX-UHFFFAOYSA-N |
Synonym | 2,4-dibromobenzene-1-sulfonyl chloride, 2,4-dibromobenzenesulphonyl chloride, 2,4-dibromophenyl chlorosulfone, pubchem5111, 2,4-dibromophenylsulfonyl chloride, benzenesulfonylchloride, 2,4-dibromo, 2,4-bis bromanyl benzenesulfonyl chloride, 2,4-dibromobenzenesulfonyl chloride |
PubChem CID | 2737366 |
IUPAC Name | 2,4-dibromobenzenesulfonyl chloride |
SMILES | ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1988703
|
Thermo Scientific Chemicals
L1988703 |
1 g |
Each for $94.71
|
|
|||||
AAL1988706
|
Thermo Scientific Chemicals
L1988706 |
5 g |
Each for $321.33
|
|
|||||
Description
2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Chemical Identifiers
72256-95-2 | |
334.41 | |
OMOXIKAWVFDFNX-UHFFFAOYSA-N | |
2737366 | |
ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1 |
C6H3Br2ClO2S | |
MFCD03093759 | |
2,4-dibromobenzene-1-sulfonyl chloride, 2,4-dibromobenzenesulphonyl chloride, 2,4-dibromophenyl chlorosulfone, pubchem5111, 2,4-dibromophenylsulfonyl chloride, benzenesulfonylchloride, 2,4-dibromo, 2,4-bis bromanyl benzenesulfonyl chloride, 2,4-dibromobenzenesulfonyl chloride | |
2,4-dibromobenzenesulfonyl chloride |
Specifications
72256-95-2 | |
128°C (0.2 mmHg) | |
MFCD03093759 | |
UN3261 | |
Moisture sensitive | |
Insoluble in water. | |
ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1 | |
334.41 | |
334.41 | |
2,4-Dibromobenzenesulfonyl chloride |
82°C to 86°C | |
C6H3Br2ClO2S | |
1 g | |
2694058 | |
2,4-dibromobenzene-1-sulfonyl chloride, 2,4-dibromobenzenesulphonyl chloride, 2,4-dibromophenyl chlorosulfone, pubchem5111, 2,4-dibromophenylsulfonyl chloride, benzenesulfonylchloride, 2,4-dibromo, 2,4-bis bromanyl benzenesulfonyl chloride, 2,4-dibromobenzenesulfonyl chloride | |
OMOXIKAWVFDFNX-UHFFFAOYSA-N | |
2,4-dibromobenzenesulfonyl chloride | |
2737366 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only