missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Difluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2382106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,4-Difluorobenzeneboronic acid | |
247°C to 250°C | |
MFCD01318998 | |
8616257 | |
QQLRSCZSKQTFGY-UHFFFAOYSA-N | |
(2,4-difluorophenyl)boronic acid | |
2734334 | |
97% |
144025-03-6 | |
C6H5BF2O2 | |
5 g | |
2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl | |
OB(O)C1=CC=C(F)C=C1F | |
157.91 | |
157.91 |
Chemical Identifiers
144025-03-6 | |
157.91 | |
QQLRSCZSKQTFGY-UHFFFAOYSA-N | |
2734334 | |
OB(O)C1=CC=C(F)C=C1F |
C6H5BF2O2 | |
MFCD01318998 | |
2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl | |
(2,4-difluorophenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only