missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Difluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

$98.62 - $460.19

Chemical Identifiers

CAS 144025-03-6
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.91
MDL Number MFCD01318998
InChI Key QQLRSCZSKQTFGY-UHFFFAOYSA-N
Synonym 2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl
PubChem CID 2734334
IUPAC Name (2,4-difluorophenyl)boronic acid
SMILES OB(O)C1=CC=C(F)C=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2382106
View Documents
Thermo Scientific Chemicals
B2382106
5 g
Each for $98.62
 
AAB2382114
View Documents
Thermo Scientific Chemicals
B2382114
25 g
Each for $460.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

144025-03-6
157.91
QQLRSCZSKQTFGY-UHFFFAOYSA-N
2734334
OB(O)C1=CC=C(F)C=C1F
C6H5BF2O2
MFCD01318998
2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl
(2,4-difluorophenyl)boronic acid

Specifications

144025-03-6
C6H5BF2O2
5 g
2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl
OB(O)C1=CC=C(F)C=C1F
157.91
157.91
2,4-Difluorobenzeneboronic acid
247°C to 250°C
MFCD01318998
8616257
QQLRSCZSKQTFGY-UHFFFAOYSA-N
(2,4-difluorophenyl)boronic acid
2734334
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only