missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[(4-Fluorophenyl)thio]acetic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 332-51-4
Molecular Formula C8H7FO2S
Molecular Weight (g/mol) 186.2
MDL Number MFCD00219825
InChI Key HQEROVXUKINLPI-UHFFFAOYSA-N
Synonym 4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid
PubChem CID 736197
IUPAC Name 2-(4-fluorophenyl)sulfanylacetic acid
SMILES C1=CC(=CC=C1F)SCC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KM09206DA
SDS View Documents
Thermo Scientific Chemicals
KM09206DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

332-51-4
186.2
HQEROVXUKINLPI-UHFFFAOYSA-N
736197
C1=CC(=CC=C1F)SCC(=O)O
C8H7FO2S
MFCD00219825
4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid
2-(4-fluorophenyl)sulfanylacetic acid

Specifications

332-51-4
97%
C8H7FO2S
MFCD00219825
4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid
C1=CC(=CC=C1F)SCC(=O)O
186.2
186.21
2-[(4-Fluorophenyl)thio]acetic acid
97.0
Amber Glass Bottle
FC6H4SCH2CO2H
1g
HQEROVXUKINLPI-UHFFFAOYSA-N
2-(4-fluorophenyl)sulfanylacetic acid
736197
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'