missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Pentanedione, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 129960025

 View more versions of this product

Catalog No. AC129960025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4-Pentanedione
-23.0°C
133.0°C to 135.0°C
Authentic
Glass bottle
1.4510 to 1.4530
MFCD00008787
01,777
0.975
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
31261
CHEBI:14750
99+%
123-54-6
0.9750g/mL
35.5°C
99% min. (GC)
C5H8O2
CH3COCH2COCH3
2.5 kg
01,1
15,76
Solubility in water: 160g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol, chloroform, ether, benzene, and acetone
CC(=O)CC(C)=O
100.12
0.7445 mPa.s (20°C)
100.12
Liquid

Chemical Identifiers

123-54-6
100.12
YRKCREAYFQTBPV-UHFFFAOYSA-N
31261
pentane-2,4-dione
C5H8O2
MFCD00008787
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
CHEBI:14750
CC(=O)CC(C)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 204-634-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only