Learn More
2,4-Pentanedione, 99+%, Thermo Scientific Chemicals
$58.75 - $245.18
Chemical Identifiers
CAS | 123-54-6 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00008787 |
InChI Key | YRKCREAYFQTBPV-UHFFFAOYSA-N |
Synonym | 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone |
PubChem CID | 31261 |
ChEBI | CHEBI:14750 |
IUPAC Name | pentane-2,4-dione |
SMILES | CC(=O)CC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC129960050
|
Thermo Scientific Chemicals
129960050 |
5 g |
Each for $58.75
|
|
|||||
AC129962500
|
Thermo Scientific Chemicals
129962500 |
250 g |
Each for $86.42
|
|
|||||
AC129960010
|
Thermo Scientific Chemicals
129960010 |
1 kg |
Each for $135.62
|
|
|||||
AC129960025
|
Thermo Scientific Chemicals
129960025 |
2.5 kg |
Each for $245.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-54-6 | |
100.12 | |
YRKCREAYFQTBPV-UHFFFAOYSA-N | |
31261 | |
pentane-2,4-dione |
C5H8O2 | |
MFCD00008787 | |
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone | |
CHEBI:14750 | |
CC(=O)CC(C)=O |
Specifications
123-54-6 | |
0.9750g/mL | |
35.5°C | |
99% min. (GC) | |
C5H8O2 | |
CH3COCH2COCH3 | |
5 g | |
01,1 | |
15,76 | |
Solubility in water: 160g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol, chloroform, ether, benzene, and acetone | |
CC(=O)CC(C)=O | |
100.12 | |
0.7445 mPa.s (20°C) | |
100.12 | |
Liquid |
-23.0°C | |
133.0°C to 135.0°C | |
Authentic | |
Glass bottle | |
1.4510 to 1.4530 | |
MFCD00008787 | |
01,777 | |
0.975 | |
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone | |
YRKCREAYFQTBPV-UHFFFAOYSA-N | |
pentane-2,4-dione | |
31261 | |
CHEBI:14750 | |
99+% | |
2, 4-Pentanedione |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 204-634-
RUO – Research Use Only