missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluorophenylacetonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 311660050

 View more versions of this product

Catalog No. AC311660050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2, 5-Difluorophenylacetonitrile
96.0
1.2300g/mL
Authentic
Glass bottle
1.4824 to 1.4844
MFCD00009972
1.23
UIMMFRUOZOWROM-UHFFFAOYSA-N
2-(2,5-difluorophenyl)acetonitrile
592780
97%
69584-87-8
100.0
102°C
96% min. (GC)
C8H5F2N
F2C6H3CH2CN
5g
2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
FC1=CC=C(F)C(CC#N)=C1
153.13
153.13

Chemical Identifiers

69584-87-8
153.13
UIMMFRUOZOWROM-UHFFFAOYSA-N
592780
FC1=CC=C(F)C(CC#N)=C1
C8H5F2N
MFCD00009972
2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
2-(2,5-difluorophenyl)acetonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 274-051-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only