missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluorophenylacetonitrile, 97%, Thermo Scientific™

Chemical Identifiers

CAS 69584-87-8
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00009972
InChI Key UIMMFRUOZOWROM-UHFFFAOYSA-N
Synonym 2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
PubChem CID 592780
IUPAC Name 2-(2,5-difluorophenyl)acetonitrile
SMILES FC1=CC=C(F)C(CC#N)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC311660050
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
311660050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

69584-87-8
153.13
UIMMFRUOZOWROM-UHFFFAOYSA-N
592780
FC1=CC=C(F)C(CC#N)=C1
C8H5F2N
MFCD00009972
2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
2-(2,5-difluorophenyl)acetonitrile

Specifications

69584-87-8
100.0
102°C
96% min. (GC)
C8H5F2N
F2C6H3CH2CN
5g
2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
FC1=CC=C(F)C(CC#N)=C1
153.13
153.13
2, 5-Difluorophenylacetonitrile
96.0
1.2300g/mL
Authentic
Glass bottle
1.4824 to 1.4844
MFCD00009972
1.23
UIMMFRUOZOWROM-UHFFFAOYSA-N
2-(2,5-difluorophenyl)acetonitrile
592780
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 274-051-4

SDS
Documents

Documents

RUO – Research Use Only