missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluorophenylboronic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 359140010

 View more versions of this product

Catalog No. AC359140010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 5-Difluorophenylboronic acid
96.0
Authentic
Glass bottle
1g
KTOJGSDLJNUAEP-UHFFFAOYSA-N
(2,5-difluorophenyl)boronic acid
2734335
97%
193353-34-3
100.0
96% min. (HPLC)
C6H5BF2O2
2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
B(C1=C(C=CC(=C1)F)F)(O)O
157.91
157.91

Chemical Identifiers

193353-34-3
157.91
2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
(2,5-difluorophenyl)boronic acid
C6H5BF2O2
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2734335
B(C1=C(C=CC(=C1)F)F)(O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only