missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluorophenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 193353-34-3
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.91
InChI Key KTOJGSDLJNUAEP-UHFFFAOYSA-N
Synonym 2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
PubChem CID 2734335
IUPAC Name (2,5-difluorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)F)F)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359140010
SDS View Documents
Thermo Scientific Chemicals
359140010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

193353-34-3
157.91
2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
(2,5-difluorophenyl)boronic acid
C6H5BF2O2
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2734335
B(C1=C(C=CC(=C1)F)F)(O)O

Specifications

193353-34-3
100.0
96% min. (HPLC)
C6H5BF2O2
2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
B(C1=C(C=CC(=C1)F)F)(O)O
157.91
157.91
2, 5-Difluorophenylboronic acid
96.0
Authentic
Glass bottle
1g
KTOJGSDLJNUAEP-UHFFFAOYSA-N
(2,5-difluorophenyl)boronic acid
2734335
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only