missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethyl-2,5-hexanediol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 162502500

 View more versions of this product

Catalog No. AC162502500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 5-Dimethyl-2, 5-hexanediol
110-03-2
GC≥96.0%
88°C to 91°C
214°C to 215°C
Conforms
Glass bottle
HOC(CH3)2CH2CH2C(CH3)2OH
250 g
2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830
ZWNMRZQYWRLGMM-UHFFFAOYSA-N
2,5-dimethylhexane-2,5-diol
8031
97%
97%
98.5
100.0
White
126°C
96% min. (GC)
C8H18O2
MFCD00004473
01, 492
Solubility in water: soluble. Other solubilities: soluble in acetone,ethylene glycol,insoluble in benzene,ccl4,petroleuw ether
CC(C)(O)CCC(C)(C)O
146.23
146.23
Flakes, Pellets or Prills

Chemical Identifiers

110-03-2
146.23
ZWNMRZQYWRLGMM-UHFFFAOYSA-N
8031
CC(C)(O)CCC(C)(C)O
C8H18O2
MFCD00004473
2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830
2,5-dimethylhexane-2,5-diol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS Signal Word: Danger

EINECSNumber : 203-731-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only