missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichloro-3-nitrotoluene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2052018

 View more versions of this product

Catalog No. AAB2052018


Explore available promotions

Description

Description

2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2,6-Dichloro-3-nitrotoluene
53°C to 55°C
MFCD00024185
UN2811
2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
WBNZUUIFTPNYRN-UHFFFAOYSA-N
1,3-dichloro-2-methyl-4-nitrobenzene
97155
99%
29682-46-0
C7H5Cl2NO2
50 g
2259855
Insoluble in water.
CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
206.02
206.03

Chemical Identifiers

29682-46-0
206.02
WBNZUUIFTPNYRN-UHFFFAOYSA-N
97155
CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
C7H5Cl2NO2
MFCD00024185
2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
1,3-dichloro-2-methyl-4-nitrobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 249-776-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only