missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichloro-3-nitrotoluene, 99%, Thermo Scientific Chemicals

$69.93 - $255.67

Chemical Identifiers

CAS 29682-46-0
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00024185
InChI Key WBNZUUIFTPNYRN-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
PubChem CID 97155
IUPAC Name 1,3-dichloro-2-methyl-4-nitrobenzene
SMILES CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2052009
View Documents
Thermo Scientific Chemicals
B2052009
10 g
Each for $69.93
 
AAB2052018
View Documents
Thermo Scientific Chemicals
B2052018
50 g
Each for $255.67
 
Description

Description

2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

29682-46-0
206.02
WBNZUUIFTPNYRN-UHFFFAOYSA-N
97155
CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
C7H5Cl2NO2
MFCD00024185
2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
1,3-dichloro-2-methyl-4-nitrobenzene

Specifications

29682-46-0
C7H5Cl2NO2
10 g
2259855
Insoluble in water.
CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
206.02
206.03
2,6-Dichloro-3-nitrotoluene
53°C to 55°C
MFCD00024185
UN2811
2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
WBNZUUIFTPNYRN-UHFFFAOYSA-N
1,3-dichloro-2-methyl-4-nitrobenzene
97155
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 249-776-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only