missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Difluoro-4-hydroxybenzaldehyde, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3197403

 View more versions of this product

Catalog No. AAH3197403

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,6-Difluoro-4-hydroxybenzaldehyde
179°C to 182°C
MFCD07772042
Air Sensitive
ROAQMGJHSNIROA-UHFFFAOYSA-N
2,6-difluoro-4-hydroxybenzaldehyde
24903548
95%
532967-21-8
C7H4F2O2
1 g
2,6-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy, ksc495q9f, 3,5-difluoro-4-formylphenol, 4-hydroxy-2,6-difluorobenzaldehyde, 2,6-difluoro-4-hydroxy benzaldehyde, 2,6-bis fluoranyl-4-oxidanyl-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy-9ci
C1=C(C=C(C(=C1F)C=O)F)O
158.104
158.11

Chemical Identifiers

532967-21-8
158.104
ROAQMGJHSNIROA-UHFFFAOYSA-N
24903548
C1=C(C=C(C(=C1F)C=O)F)O
C7H4F2O2
MFCD07772042
2,6-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy, ksc495q9f, 3,5-difluoro-4-formylphenol, 4-hydroxy-2,6-difluorobenzaldehyde, 2,6-difluoro-4-hydroxy benzaldehyde, 2,6-bis fluoranyl-4-oxidanyl-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy-9ci
2,6-difluoro-4-hydroxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only