missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Difluoro-4-hydroxybenzaldehyde, 95%, Thermo Scientific Chemicals

$109.58 - $160.53

Chemical Identifiers

CAS 532967-21-8
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD07772042
InChI Key ROAQMGJHSNIROA-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy, ksc495q9f, 3,5-difluoro-4-formylphenol, 4-hydroxy-2,6-difluorobenzaldehyde, 2,6-difluoro-4-hydroxy benzaldehyde, 2,6-bis fluoranyl-4-oxidanyl-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy-9ci
PubChem CID 24903548
IUPAC Name 2,6-difluoro-4-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1F)C=O)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH31974MD
View Documents
Thermo Scientific Chemicals
H31974MD
250 mg
Each for $109.58
 
AAH3197403
View Documents
Thermo Scientific Chemicals
H3197403
1 g
Each for $160.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

532967-21-8
158.104
ROAQMGJHSNIROA-UHFFFAOYSA-N
24903548
C1=C(C=C(C(=C1F)C=O)F)O
C7H4F2O2
MFCD07772042
2,6-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy, ksc495q9f, 3,5-difluoro-4-formylphenol, 4-hydroxy-2,6-difluorobenzaldehyde, 2,6-difluoro-4-hydroxy benzaldehyde, 2,6-bis fluoranyl-4-oxidanyl-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy-9ci
2,6-difluoro-4-hydroxybenzaldehyde

Specifications

532967-21-8
C7H4F2O2
250 mg
2,6-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy, ksc495q9f, 3,5-difluoro-4-formylphenol, 4-hydroxy-2,6-difluorobenzaldehyde, 2,6-difluoro-4-hydroxy benzaldehyde, 2,6-bis fluoranyl-4-oxidanyl-benzaldehyde, benzaldehyde, 2,6-difluoro-4-hydroxy-9ci
C1=C(C=C(C(=C1F)C=O)F)O
158.104
158.11
2,6-Difluoro-4-hydroxybenzaldehyde
179°C to 182°C
MFCD07772042
Air Sensitive
ROAQMGJHSNIROA-UHFFFAOYSA-N
2,6-difluoro-4-hydroxybenzaldehyde
24903548
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only