missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 116570010

 View more versions of this product

Catalog No. AC116570010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 6-Dimethylphenol
98.5
45.0°C to 48.0°C
1.1500g/mL
73°C
98.5% min. (GC)
C8H10O
MFCD00002240
06, 485
15, 10279
Solubility in water: 10g/L (20°C). Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene
CC1=CC=CC(C)=C1O
122.17
122.17
Low Melting Solid
576-26-1
100.0
Brown-Yellow to Colorless
203.0°C
Authentic
Glass bottle
(CH3)2C6H3OH
1 kg
1.15
2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene
NXXYKOUNUYWIHA-UHFFFAOYSA-N
2,6-dimethylphenol
11335
99%

Chemical Identifiers

576-26-1
122.17
NXXYKOUNUYWIHA-UHFFFAOYSA-N
11335
CC1=CC=CC(C)=C1O
C8H10O
MFCD00002240
2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene
2,6-dimethylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-400-1

Recommended Storage : this product gradually takes up color on storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only