missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals

$32.02 - $134.11

Chemical Identifiers

CAS 576-26-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002240
InChI Key NXXYKOUNUYWIHA-UHFFFAOYSA-N
Synonym 2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene
PubChem CID 11335
IUPAC Name 2,6-dimethylphenol
SMILES CC1=CC=CC(C)=C1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC116570050
SDS View Documents
Thermo Scientific Chemicals
116570050
5 g Glass bottle
Each for $32.02
 
AC116572500
SDS View Documents
Thermo Scientific Chemicals
116572500
250 g Glass bottle
Each for $51.04
 
AC116570010
SDS View Documents
Thermo Scientific Chemicals
116570010
1 kg Glass bottle
Each for $134.11
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

576-26-1
122.17
NXXYKOUNUYWIHA-UHFFFAOYSA-N
11335
CC1=CC=CC(C)=C1O
C8H10O
MFCD00002240
2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene
2,6-dimethylphenol

Specifications

576-26-1
100.0
Brown-Yellow to Colorless
203.0°C
Authentic
Glass bottle
(CH3)2C6H3OH
5 g
1.15
2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene
NXXYKOUNUYWIHA-UHFFFAOYSA-N
2,6-dimethylphenol
11335
99%
2, 6-Dimethylphenol
98.5
45.0°C to 48.0°C
1.1500g/mL
73°C
98.5% min. (GC)
C8H10O
MFCD00002240
06, 485
15, 10279
Solubility in water: 10g/L (20°C). Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene
CC1=CC=CC(C)=C1O
122.17
122.17
Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-400-1

Recommended Storage : this product gradually takes up color on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only