missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetylfuran, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1493818

 View more versions of this product

Catalog No. AAA1493818


Explore available promotions

Description

Description

2-Acetylfuran is a useful intermediate in the synthesis of fine chemicals and pharmaceuticals, and is used in the production of the generic cephalophosphorin antibiotic cefuroxime

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylfuran is a useful intermediate in the synthesis of fine chemicals and pharmaceuticals, and is used in the production of the generic cephalophosphorin antibiotic cefuroxime

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Acetylfuran
27°C to 30°C
171°C to 173°C
C6H6O2
MFCD00003242
UN2811
2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanyl
IEMMBWWQXVXBEU-UHFFFAOYSA-N
1-(furan-2-yl)ethanone
14505
110.11
1192-62-7
1.098
71°C (159°F)
1.507
50 g
107909
Insoluble in water.
CC(=O)C1=CC=CO1
110.112
CHEBI:59983
99%

Chemical Identifiers

1192-62-7
110.112
IEMMBWWQXVXBEU-UHFFFAOYSA-N
14505
1-(furan-2-yl)ethanone
C6H6O2
MFCD00003242
2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanyl
CHEBI:59983
CC(=O)C1=CC=CO1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H312-H332-H319
Fatal if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 214-757-1

RTECSNumber : OB3870000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only