missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetylfuran, 99%, Thermo Scientific Chemicals

$70.30 - $186.27

Chemical Identifiers

CAS 1192-62-7
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00003242
InChI Key IEMMBWWQXVXBEU-UHFFFAOYSA-N
Synonym 2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanyl
PubChem CID 14505
ChEBI CHEBI:59983
IUPAC Name 1-(furan-2-yl)ethanone
SMILES CC(=O)C1=CC=CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1493818
View Documents
Thermo Scientific Chemicals
A1493818
50 g
Each for $70.30
 
AAA1493830
View Documents
Thermo Scientific Chemicals
A1493830
250 g
Each for $186.27
 
Description

Description

2-Acetylfuran is a useful intermediate in the synthesis of fine chemicals and pharmaceuticals, and is used in the production of the generic cephalophosphorin antibiotic cefuroxime

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylfuran is a useful intermediate in the synthesis of fine chemicals and pharmaceuticals, and is used in the production of the generic cephalophosphorin antibiotic cefuroxime

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

1192-62-7
110.112
IEMMBWWQXVXBEU-UHFFFAOYSA-N
14505
1-(furan-2-yl)ethanone
C6H6O2
MFCD00003242
2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanyl
CHEBI:59983
CC(=O)C1=CC=CO1

Specifications

1192-62-7
1.098
71°C (159°F)
1.507
50 g
107909
Insoluble in water.
CC(=O)C1=CC=CO1
110.112
CHEBI:59983
99%
27°C to 30°C
171°C to 173°C
C6H6O2
MFCD00003242
UN2811
2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanyl
IEMMBWWQXVXBEU-UHFFFAOYSA-N
1-(furan-2-yl)ethanone
14505
110.11
2-Acetylfuran
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H312-H332-H319
Fatal if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 214-757-1

RTECSNumber : OB3870000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only