missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Allylphenol, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1075536

 View more versions of this product

Catalog No. AAA1075536


Explore available promotions

Description

Description

2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.

Solubility
Soluble in water(7 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications

Specifications

2-Allylphenol
-6°C
220°C to 222°C
Phenol-like
1.545
500 g
742121
Soluble in water(7g/L).
C=CCC1=CC=CC=C1O
134.178
CHEBI:39826
≥98%
1745-81-9
1.024
88°C (190°F)
C9H10O
MFCD00002250
UN2922
2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol
QIRNGVVZBINFMX-UHFFFAOYSA-N
2-prop-2-enylphenol
15624
134.18

Chemical Identifiers

1745-81-9
134.178
QIRNGVVZBINFMX-UHFFFAOYSA-N
15624
2-prop-2-enylphenol
C9H10O
MFCD00002250
2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol
CHEBI:39826
C=CCC1=CC=CC=C1O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302+H312-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 217-119-0

RTECSNumber : SJ3850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only