Learn More
2-Allylphenol, 98+%, Thermo Scientific Chemicals
$88.40 - $277.19
Chemical Identifiers
CAS | 1745-81-9 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.178 |
MDL Number | MFCD00002250 |
InChI Key | QIRNGVVZBINFMX-UHFFFAOYSA-N |
Synonym | 2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol |
PubChem CID | 15624 |
ChEBI | CHEBI:39826 |
IUPAC Name | 2-prop-2-enylphenol |
SMILES | C=CCC1=CC=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1075522
|
Thermo Scientific Chemicals
A1075522 |
100 g |
Each for $88.40
|
|
|||||
AAA1075536
|
Thermo Scientific Chemicals
A1075536 |
500 g |
Each for $277.19
|
|
|||||
Description
2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Allylphenol is an protected intermediate in the synthesis of metabolites of Diclofenac (D436450), a nonsteroidal anti-inflammatory compound an decycloxygenase (COX) inhibitor.
Solubility
Soluble in water(7 g/L).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Chemical Identifiers
1745-81-9 | |
134.178 | |
QIRNGVVZBINFMX-UHFFFAOYSA-N | |
15624 | |
2-prop-2-enylphenol |
C9H10O | |
MFCD00002250 | |
2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol | |
CHEBI:39826 | |
C=CCC1=CC=CC=C1O |
Specifications
1745-81-9 | |
1.024 | |
88°C (190°F) | |
C9H10O | |
MFCD00002250 | |
UN2922 | |
2-allylphenol, o-allylphenol, phenol, o-allyl, phenol, 2-2-propenyl, phenol, 2-propenyl, 2-prop-2-en-1-yl phenol, 2-2-propenyl phenol, 2-allyl-phenol, unii-o04f145zjz, 2-prop-2-en-1-ylphenol | |
QIRNGVVZBINFMX-UHFFFAOYSA-N | |
2-prop-2-enylphenol | |
15624 | |
134.18 | |
2-Allylphenol |
-6°C | |
220°C to 222°C | |
Phenol-like | |
1.545 | |
100 g | |
742121 | |
Soluble in water(7g/L). | |
C=CCC1=CC=CC=C1O | |
134.178 | |
CHEBI:39826 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 217-119-0
RTECSNumber : SJ3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only