missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-1,3,5-triazine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 295240050

 View more versions of this product

Catalog No. AC295240050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-1,3,5-triazine
97.5
219.0°C to 224.0°C
Authentic
Glass bottle
5 g
KCZIUKYAJJEIQG-UHFFFAOYSA-N
[diacetyloxy(ethenyl)silyl] acetate
77780
98%
4122-04-7
100.0
White to Yellow
98%
C3H4N4
vinyltriacetoxysilane, triacetoxyvinylsilane, triacetoxy vinyl silane, vinylsilanetriyl triacetate, ethenylsilanetriyl triacetate, vinyl triacetoxy silane, vinyltriacetoxy-silane, silanetriol, ethenyl-, triacetate, vinylsilanetriol triacetate, unii-w49sgj3f9r
NC1=NC=NC=N1
96.09
96.09
Crystalline Powder

Chemical Identifiers

4122-04-7
C3H4N4
96.09
KCZIUKYAJJEIQG-UHFFFAOYSA-N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 223-920-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only