missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-1,3,5-triazine, 98%, Thermo Scientific Chemicals

$146.72 - $360.53

Chemical Identifiers

CAS 4122-04-7
Molecular Formula C3H4N4
Molecular Weight (g/mol) 96.09
InChI Key KCZIUKYAJJEIQG-UHFFFAOYSA-N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC295240010
SDS View Documents
Thermo Scientific Chemicals
295240010
1 g Glass bottle
Each for $146.72
 
AC295240050
SDS View Documents
Thermo Scientific Chemicals
295240050
5 g Glass bottle
Each for $360.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4122-04-7
C3H4N4
96.09
KCZIUKYAJJEIQG-UHFFFAOYSA-N

Specifications

4122-04-7
100.0
White to Yellow
98%
C3H4N4
2-amino-1,3,5-triazine, amino-1,3,5-triazine, 2-amino-s-triazine, 1,3,5-triazine-2-ylamine, chembl89436, amino-s-triazine, s-triazine-2-amine, pubchem14498, 1,3,5 triazin-2-ylamine
C1=NC=NC(=N1)N
96.09
96.09
Crystalline Powder
97.5
219.0°C to 224.0°C
Authentic
Glass bottle
1 g
KCZIUKYAJJEIQG-UHFFFAOYSA-N
1,3,5-triazin-2-amine
20077
98%
2-Amino-1,3,5-triazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 223-920-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only