missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-2-methyl-1-propanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 104060050

 View more versions of this product

Catalog No. AC104060050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-2-methyl-1-propanol
98.5
31.0°C to 32.0°C
0.9300g/mL
67°C
99%
C4H11NO
(CH3)2C(NH2)CH2OH
5 L
01,37; 03,14; 06,20
15, 446
Solubility in water: miscible. Other solubilities: soluble in alcohols
CC(C)(N)CO
89.14
102 mPa.s (30°C)
99%
124-68-5
100.0
Colorless
165.0°C
Authentic
Plastic drum
1.4470 to 1.4490
MFCD00008051
04, III, 783
0.93
2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
CBTVGIZVANVGBH-UHFFFAOYSA-N
2-amino-2-methylpropan-1-ol
11807
89.14
Low Melting Solid

Chemical Identifiers

124-68-5
89.14
CBTVGIZVANVGBH-UHFFFAOYSA-N
11807
CC(C)(N)CO
C4H11NO
MFCD00008051
2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
2-amino-2-methylpropan-1-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-709-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only