Learn More
2-Amino-2-methyl-1-propanol, 99%, Thermo Scientific Chemicals
$70.26 - $390.53
Chemical Identifiers
CAS | 124-68-5 |
---|---|
Molecular Formula | C4H11NO |
Molecular Weight (g/mol) | 89.14 |
MDL Number | MFCD00008051 |
InChI Key | CBTVGIZVANVGBH-UHFFFAOYSA-N |
Synonym | 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine |
PubChem CID | 11807 |
IUPAC Name | 2-amino-2-methylpropan-1-ol |
SMILES | CC(C)(N)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC104065000
|
Thermo Scientific Chemicals
104065000 |
500 mL | Glass bottle |
N/A
|
|
||||
AC104060010
|
Thermo Scientific Chemicals
104060010 |
1 L | Glass bottle |
N/A
|
|
||||
AC104060050
|
Thermo Scientific Chemicals
104060050 |
5 L | Plastic drum |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-68-5 | |
89.14 | |
CBTVGIZVANVGBH-UHFFFAOYSA-N | |
11807 | |
CC(C)(N)CO |
C4H11NO | |
MFCD00008051 | |
2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine | |
2-amino-2-methylpropan-1-ol |
Specifications
124-68-5 | |
100.0 | |
Colorless | |
165.0°C | |
Authentic | |
Glass bottle | |
1.4470 to 1.4490 | |
MFCD00008051 | |
04, III, 783 | |
0.93 | |
2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine | |
CBTVGIZVANVGBH-UHFFFAOYSA-N | |
2-amino-2-methylpropan-1-ol | |
11807 | |
89.14 | |
Low Melting Solid |
98.5 | |
31.0°C to 32.0°C | |
0.9300g/mL | |
67°C | |
99% | |
C4H11NO | |
(CH3)2C(NH2)CH2OH | |
500 mL | |
01,37; 03,14; 06,20 | |
15, 446 | |
Solubility in water: miscible. Other solubilities: soluble in alcohols | |
CC(C)(N)CO | |
89.14 | |
102 mPa.s (30°C) | |
99% | |
2-Amino-2-methyl-1-propanol |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-709-8
RUO – Research Use Only