missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-3-chloro-5-trifluoromethylpyridine 98.0+%, TCI America™
Supplier: TCI America A115725G
Specifications
2-Amino-3-chloro-5-trifluoromethylpyridine | |
96°C | |
MFCD00042154 | |
3-chloro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-chloro-5-trifluoromethyl pyridine, 2-amino-3-chloro-5-trifluoromethylpyridine, 3-chloro-5-trifluoromethyl-2-pyridinamine, 3-chloro-5-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridin-2-ylamine, 2-amino-3-chloro-5-trifluoropyridine, pubchem2974 | |
NC1=C(Cl)C=C(C=N1)C(F)(F)F | |
196.56 | |
196.56 | |
Crystalline Powder |
79456-26-1 | |
C6H4ClF3N2 | |
25 g | |
WXNPZQIRDCDLJD-UHFFFAOYSA-N | |
3-chloro-5-(trifluoromethyl)pyridin-2-amine | |
607248 | |
≥98.0% (GC) |
Chemical Identifiers
79456-26-1 | |
196.56 | |
WXNPZQIRDCDLJD-UHFFFAOYSA-N | |
607248 | |
NC1=C(Cl)C=C(C=N1)C(F)(F)F |
C6H4ClF3N2 | |
MFCD00042154 | |
3-chloro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-chloro-5-trifluoromethyl pyridine, 2-amino-3-chloro-5-trifluoromethylpyridine, 3-chloro-5-trifluoromethyl-2-pyridinamine, 3-chloro-5-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridin-2-ylamine, 2-amino-3-chloro-5-trifluoropyridine, pubchem2974 | |
3-chloro-5-(trifluoromethyl)pyridin-2-amine |
Safety and Handling
TSCA : No