missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-chloro-5-trifluoromethylpyridine 98.0+%, TCI America™
$70.84 - $208.78
Chemical Identifiers
| CAS | 79456-26-1 |
|---|---|
| Molecular Formula | C6H4ClF3N2 |
| Molecular Weight (g/mol) | 196.56 |
| MDL Number | MFCD00042154 |
| InChI Key | WXNPZQIRDCDLJD-UHFFFAOYSA-N |
| Synonym | 3-chloro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-chloro-5-trifluoromethyl pyridine, 2-amino-3-chloro-5-trifluoromethylpyridine, 3-chloro-5-trifluoromethyl-2-pyridinamine, 3-chloro-5-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridin-2-ylamine, 2-amino-3-chloro-5-trifluoropyridine, pubchem2974 |
| PubChem CID | 607248 |
| IUPAC Name | 3-chloro-5-(trifluoromethyl)pyridin-2-amine |
| SMILES | NC1=C(Cl)C=C(C=N1)C(F)(F)F |
Chemical Identifiers
| 79456-26-1 | |
| 196.56 | |
| WXNPZQIRDCDLJD-UHFFFAOYSA-N | |
| 607248 | |
| NC1=C(Cl)C=C(C=N1)C(F)(F)F |
| C6H4ClF3N2 | |
| MFCD00042154 | |
| 3-chloro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-chloro-5-trifluoromethyl pyridine, 2-amino-3-chloro-5-trifluoromethylpyridine, 3-chloro-5-trifluoromethyl-2-pyridinamine, 3-chloro-5-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridin-2-ylamine, 2-amino-3-chloro-5-trifluoropyridine, pubchem2974 | |
| 3-chloro-5-(trifluoromethyl)pyridin-2-amine |
Specifications
| 79456-26-1 | |
| C6H4ClF3N2 | |
| 5 g | |
| WXNPZQIRDCDLJD-UHFFFAOYSA-N | |
| 3-chloro-5-(trifluoromethyl)pyridin-2-amine | |
| 607248 | |
| ≥98.0% (GC) | |
| 2-Amino-3-chloro-5-trifluoromethylpyridine |
| 96°C | |
| MFCD00042154 | |
| 3-chloro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-chloro-5-trifluoromethyl pyridine, 2-amino-3-chloro-5-trifluoromethylpyridine, 3-chloro-5-trifluoromethyl-2-pyridinamine, 3-chloro-5-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridin-2-ylamine, 2-amino-3-chloro-5-trifluoropyridine, pubchem2974 | |
| NC1=C(Cl)C=C(C=N1)C(F)(F)F | |
| 196.56 | |
| 196.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No