Learn More
2-Amino-4-chloro-3-nitropyridine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H64678MD
Description
2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in dimethyl formamide, dimethyl sulfoxide, hot ethanol, ethyl acetate and hot methanol.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
2-Amino-4-chloro-3-nitropyridine | |
C5H4ClN3O2 | |
250 mg | |
2-amino-4-chloro-3-nitropyridine, 2-amino-3-nitro-4-chloropyridine, 4-chloro-3-nitro-2-pyridinamine, 4-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro-3-nitro, 4-chloro-3-nitro-pyridin-2-amine, pubchem17739, ksc352m6b, 3-nitro-4-chloropyridine-2-amine, 4-chloro-2-amino-3-nitropyridine | |
DIRINUVNYFAWQF-UHFFFAOYSA-N | |
4-chloro-3-nitropyridin-2-amine | |
4071694 | |
95% |
6980-08-1 | |
MFCD04116097 | |
391939 | |
Soluble in dimethyl formamide,dimethyl sulfoxide,hot ethanol,ethyl acetate and hot methanol. | |
C1=CN=C(C(=C1Cl)[N+](=O)[O-])N | |
173.56 | |
173.56 |
Chemical Identifiers
6980-08-1 | |
173.56 | |
DIRINUVNYFAWQF-UHFFFAOYSA-N | |
4071694 | |
C1=CN=C(C(=C1Cl)[N+](=O)[O-])N |
C5H4ClN3O2 | |
MFCD04116097 | |
2-amino-4-chloro-3-nitropyridine, 2-amino-3-nitro-4-chloropyridine, 4-chloro-3-nitro-2-pyridinamine, 4-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro-3-nitro, 4-chloro-3-nitro-pyridin-2-amine, pubchem17739, ksc352m6b, 3-nitro-4-chloropyridine-2-amine, 4-chloro-2-amino-3-nitropyridine | |
4-chloro-3-nitropyridin-2-amine |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only