Learn More
2-Amino-4-chloro-3-nitropyridine, 95%, Thermo Scientific Chemicals
$60.97 - $558.36
Chemical Identifiers
CAS | 6980-08-1 |
---|---|
Molecular Formula | C5H4ClN3O2 |
Molecular Weight (g/mol) | 173.56 |
MDL Number | MFCD04116097 |
InChI Key | DIRINUVNYFAWQF-UHFFFAOYSA-N |
Synonym | 2-amino-4-chloro-3-nitropyridine, 2-amino-3-nitro-4-chloropyridine, 4-chloro-3-nitro-2-pyridinamine, 4-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro-3-nitro, 4-chloro-3-nitro-pyridin-2-amine, pubchem17739, ksc352m6b, 3-nitro-4-chloropyridine-2-amine, 4-chloro-2-amino-3-nitropyridine |
PubChem CID | 4071694 |
IUPAC Name | 4-chloro-3-nitropyridin-2-amine |
SMILES | C1=CN=C(C(=C1Cl)[N+](=O)[O-])N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH64678MD
|
Thermo Scientific Chemicals
H64678MD |
250 mg |
Each for $60.97
|
|
|||||
AAH6467803
|
Thermo Scientific Chemicals
H6467803 |
1 g |
Each for $153.49
|
|
|||||
AAH6467806
|
Thermo Scientific Chemicals
H6467806 |
5 g |
Each for $558.36
|
|
|||||
Description
2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in dimethyl formamide, dimethyl sulfoxide, hot ethanol, ethyl acetate and hot methanol.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Chemical Identifiers
6980-08-1 | |
173.56 | |
DIRINUVNYFAWQF-UHFFFAOYSA-N | |
4071694 | |
C1=CN=C(C(=C1Cl)[N+](=O)[O-])N |
C5H4ClN3O2 | |
MFCD04116097 | |
2-amino-4-chloro-3-nitropyridine, 2-amino-3-nitro-4-chloropyridine, 4-chloro-3-nitro-2-pyridinamine, 4-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro-3-nitro, 4-chloro-3-nitro-pyridin-2-amine, pubchem17739, ksc352m6b, 3-nitro-4-chloropyridine-2-amine, 4-chloro-2-amino-3-nitropyridine | |
4-chloro-3-nitropyridin-2-amine |
Specifications
6980-08-1 | |
MFCD04116097 | |
391939 | |
Soluble in dimethyl formamide,dimethyl sulfoxide,hot ethanol,ethyl acetate and hot methanol. | |
C1=CN=C(C(=C1Cl)[N+](=O)[O-])N | |
173.56 | |
173.56 | |
2-Amino-4-chloro-3-nitropyridine |
C5H4ClN3O2 | |
250 mg | |
2-amino-4-chloro-3-nitropyridine, 2-amino-3-nitro-4-chloropyridine, 4-chloro-3-nitro-2-pyridinamine, 4-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro-3-nitro, 4-chloro-3-nitro-pyridin-2-amine, pubchem17739, ksc352m6b, 3-nitro-4-chloropyridine-2-amine, 4-chloro-2-amino-3-nitropyridine | |
DIRINUVNYFAWQF-UHFFFAOYSA-N | |
4-chloro-3-nitropyridin-2-amine | |
4071694 | |
95% |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only