missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-chlorobenzamide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1115306

 View more versions of this product

Catalog No. AAA1115306


Explore available promotions

Description

Description

2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5'-chloro-2'-carboxamidobenzanilide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5′-chloro-2′-carboxamidobenzanilide.

Solubility
Soluble in acetone.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Amino-5-chlorobenzamide
169°C to 173°C
MFCD00017126
743347
Soluble in acetone.
C1=CC(=C(C=C1Cl)C(=O)N)N
170.596
170.6
5202-85-7
C7H7ClN2O
5 g
5-chloroanthranilamide, benzamide, 2-amino-5-chloro, 2-amino-5-chloro-benzamide, 5-chloro-2-aminobenzamide, 2-azanyl-5-chloranyl-benzamide, pubchem13624, acmc-1axdu, 3-14-00-00963 beilstein handbook reference, 5mk
DNRVZOZGQHHDAT-UHFFFAOYSA-N
2-amino-5-chlorobenzamide
78876
≥98%

Chemical Identifiers

5202-85-7
170.596
DNRVZOZGQHHDAT-UHFFFAOYSA-N
78876
C1=CC(=C(C=C1Cl)C(=O)N)N
C7H7ClN2O
MFCD00017126
5-chloroanthranilamide, benzamide, 2-amino-5-chloro, 2-amino-5-chloro-benzamide, 5-chloro-2-aminobenzamide, 2-azanyl-5-chloranyl-benzamide, pubchem13624, acmc-1axdu, 3-14-00-00963 beilstein handbook reference, 5mk
2-amino-5-chlorobenzamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-990-3

RTECSNumber : CU9090000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only