Learn More
2-Amino-5-chlorobenzamide, 98+%, Thermo Scientific Chemicals
$38.36 - $128.21
Chemical Identifiers
CAS | 5202-85-7 |
---|---|
Molecular Formula | C7H7ClN2O |
Molecular Weight (g/mol) | 170.596 |
MDL Number | MFCD00017126 |
InChI Key | DNRVZOZGQHHDAT-UHFFFAOYSA-N |
Synonym | 5-chloroanthranilamide, benzamide, 2-amino-5-chloro, 2-amino-5-chloro-benzamide, 5-chloro-2-aminobenzamide, 2-azanyl-5-chloranyl-benzamide, pubchem13624, acmc-1axdu, 3-14-00-00963 beilstein handbook reference, 5mk |
PubChem CID | 78876 |
IUPAC Name | 2-amino-5-chlorobenzamide |
SMILES | C1=CC(=C(C=C1Cl)C(=O)N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1115303
|
Thermo Scientific Chemicals
A1115303 |
1 g |
Each for $38.36
|
|
|||||
AAA1115306
|
Thermo Scientific Chemicals
A1115306 |
5 g |
Each for $128.21
|
|
|||||
Description
2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5'-chloro-2'-carboxamidobenzanilide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5′-chloro-2′-carboxamidobenzanilide.
Solubility
Soluble in acetone.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
5202-85-7 | |
170.596 | |
DNRVZOZGQHHDAT-UHFFFAOYSA-N | |
78876 | |
C1=CC(=C(C=C1Cl)C(=O)N)N |
C7H7ClN2O | |
MFCD00017126 | |
5-chloroanthranilamide, benzamide, 2-amino-5-chloro, 2-amino-5-chloro-benzamide, 5-chloro-2-aminobenzamide, 2-azanyl-5-chloranyl-benzamide, pubchem13624, acmc-1axdu, 3-14-00-00963 beilstein handbook reference, 5mk | |
2-amino-5-chlorobenzamide |
Specifications
5202-85-7 | |
C7H7ClN2O | |
1 g | |
5-chloroanthranilamide, benzamide, 2-amino-5-chloro, 2-amino-5-chloro-benzamide, 5-chloro-2-aminobenzamide, 2-azanyl-5-chloranyl-benzamide, pubchem13624, acmc-1axdu, 3-14-00-00963 beilstein handbook reference, 5mk | |
DNRVZOZGQHHDAT-UHFFFAOYSA-N | |
2-amino-5-chlorobenzamide | |
78876 | |
≥98% |
169°C to 173°C | |
MFCD00017126 | |
743347 | |
Soluble in acetone. | |
C1=CC(=C(C=C1Cl)C(=O)N)N | |
170.596 | |
170.6 | |
2-Amino-5-chlorobenzamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 225-990-3
RTECSNumber : CU9090000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only