missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-bromobenzoic acid, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3210606

 View more versions of this product

Catalog No. AAH3210606


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-6-bromobenzoic acid
C7H6BrNO2
5 g
6-bromoanthranilic acid, 3-bromo-2-carboxyaniline, benzoic acid, 2-amino-6-bromo, buttpark 49\07-50, 2-amino-6-bromo-benzoic acid, ksc201s8r
NC1=C(C(O)=O)C(Br)=CC=C1
216.03
216.03
20776-48-1
MFCD03618455
Light sensitive
BNQPROAXWQCNKO-UHFFFAOYSA-N
2-amino-6-bromobenzoic acid
10560649
98%

Chemical Identifiers

20776-48-1
216.03
BNQPROAXWQCNKO-UHFFFAOYSA-N
10560649
NC1=C(C(O)=O)C(Br)=CC=C1
C7H6BrNO2
MFCD03618455
6-bromoanthranilic acid, 3-bromo-2-carboxyaniline, benzoic acid, 2-amino-6-bromo, buttpark 49\07-50, 2-amino-6-bromo-benzoic acid, ksc201s8r
2-amino-6-bromobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only