missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-6-bromobenzoic acid, 97+%, Thermo Scientific Chemicals
$245.25 - $808.75
Chemical Identifiers
CAS | 20776-48-1 |
---|---|
Molecular Formula | C7H6BrNO2 |
Molecular Weight (g/mol) | 216.03 |
MDL Number | MFCD03618455 |
InChI Key | BNQPROAXWQCNKO-UHFFFAOYSA-N |
Synonym | 6-bromoanthranilic acid, 3-bromo-2-carboxyaniline, benzoic acid, 2-amino-6-bromo, buttpark 49\07-50, 2-amino-6-bromo-benzoic acid, ksc201s8r |
PubChem CID | 10560649 |
IUPAC Name | 2-amino-6-bromobenzoic acid |
SMILES | NC1=C(C(O)=O)C(Br)=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3210603
|
Thermo Scientific Chemicals
H3210603 |
1 g |
Each for $245.25
|
|
|||||
AAH3210606
|
Thermo Scientific Chemicals
H3210606 |
5 g |
Each for $808.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20776-48-1 | |
216.03 | |
BNQPROAXWQCNKO-UHFFFAOYSA-N | |
10560649 | |
NC1=C(C(O)=O)C(Br)=CC=C1 |
C7H6BrNO2 | |
MFCD03618455 | |
6-bromoanthranilic acid, 3-bromo-2-carboxyaniline, benzoic acid, 2-amino-6-bromo, buttpark 49\07-50, 2-amino-6-bromo-benzoic acid, ksc201s8r | |
2-amino-6-bromobenzoic acid |
Specifications
20776-48-1 | |
MFCD03618455 | |
Light sensitive | |
BNQPROAXWQCNKO-UHFFFAOYSA-N | |
2-amino-6-bromobenzoic acid | |
10560649 | |
98% |
C7H6BrNO2 | |
1 g | |
6-bromoanthranilic acid, 3-bromo-2-carboxyaniline, benzoic acid, 2-amino-6-bromo, buttpark 49\07-50, 2-amino-6-bromo-benzoic acid, ksc201s8r | |
NC1=C(C(O)=O)C(Br)=CC=C1 | |
216.03 | |
216.03 | |
2-Amino-6-bromobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only