missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminoacetophenone hydrochloride, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 103100010

 View more versions of this product

Catalog No. AC103100010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Aminoacetophenone hydrochloride
95.0
194.0°C
Authentic
Glass bottle
C6H5COCH2NH2·HCl
1 g
10, 7068
CVXGFPPAIUELDV-UHFFFAOYSA-N
2-amino-1-phenylethanone;hydrochloride
2723597
96%
5468-37-1
100.0
Yellow to Beige
95.0 to 105% (Argentometry)
C8H10ClNO
MFCD00012873
14, 49
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride
Cl.NCC(=O)C1=CC=CC=C1
171.62
171.63
Crystals

Chemical Identifiers

5468-37-1
171.62
CVXGFPPAIUELDV-UHFFFAOYSA-N
2723597
Cl.NCC(=O)C1=CC=CC=C1
C8H10ClNO
MFCD00012873
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride
2-amino-1-phenylethanone;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irri

GHS Signal Word: Warning

EINECSNumber : 226-787-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only