Learn More
2-Aminoacetophenone hydrochloride, 96%, Thermo Scientific Chemicals
$116.59 - $361.49
Chemical Identifiers
CAS | 5468-37-1 |
---|---|
Molecular Formula | C8H10ClNO |
Molecular Weight (g/mol) | 171.62 |
MDL Number | MFCD00012873 |
InChI Key | CVXGFPPAIUELDV-UHFFFAOYSA-N |
Synonym | 2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride |
PubChem CID | 2723597 |
IUPAC Name | 2-amino-1-phenylethanone;hydrochloride |
SMILES | Cl.NCC(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC103100010
|
Thermo Scientific Chemicals
103100010 |
1 g | Glass bottle |
Each for $116.59
|
|
||||
AC103100050
|
Thermo Scientific Chemicals
103100050 |
5 g | Glass bottle |
Each for $361.49
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5468-37-1 | |
171.62 | |
CVXGFPPAIUELDV-UHFFFAOYSA-N | |
2723597 | |
Cl.NCC(=O)C1=CC=CC=C1 |
C8H10ClNO | |
MFCD00012873 | |
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride | |
2-amino-1-phenylethanone;hydrochloride |
Specifications
5468-37-1 | |
100.0 | |
Yellow to Beige | |
95.0 to 105% (Argentometry) | |
C8H10ClNO | |
MFCD00012873 | |
14, 49 | |
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride | |
Cl.NCC(=O)C1=CC=CC=C1 | |
171.62 | |
171.63 | |
Crystals |
95.0 | |
194.0°C | |
Authentic | |
Glass bottle | |
C6H5COCH2NH2·HCl | |
1 g | |
10, 7068 | |
CVXGFPPAIUELDV-UHFFFAOYSA-N | |
2-amino-1-phenylethanone;hydrochloride | |
2723597 | |
96% | |
2-Aminoacetophenone hydrochloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irri
GHS Signal Word: Warning
EINECSNumber : 226-787-2
RUO – Research Use Only