missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminoacetophenone hydrochloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2911709

 View more versions of this product

Catalog No. AAH2911709

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Aminoacetophenone hydrochloride
∽194°C (decomposition)
C8H10ClNO
10 g
CVXGFPPAIUELDV-UHFFFAOYSA-N
2-amino-1-phenylethanone;hydrochloride
2723597
98%
5468-37-1
Odorless
MFCD00012873
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride
Cl.NCC(=O)C1=CC=CC=C1
171.62
171.63

Chemical Identifiers

5468-37-1
171.62
CVXGFPPAIUELDV-UHFFFAOYSA-N
2723597
Cl.NCC(=O)C1=CC=CC=C1
C8H10ClNO
MFCD00012873
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride
2-amino-1-phenylethanone;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

EINECSNumber : 226-787-2

RTECSNumber : AM5940000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only