Learn More
2-Aminoacetophenone hydrochloride, 97%, Thermo Scientific Chemicals
$94.05 - $420.03
Chemical Identifiers
CAS | 5468-37-1 |
---|---|
Molecular Formula | C8H10ClNO |
Molecular Weight (g/mol) | 171.62 |
MDL Number | MFCD00012873 |
InChI Key | CVXGFPPAIUELDV-UHFFFAOYSA-N |
Synonym | 2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride |
PubChem CID | 2723597 |
IUPAC Name | 2-amino-1-phenylethanone;hydrochloride |
SMILES | Cl.NCC(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH2911703
|
Thermo Scientific Chemicals
H2911703 |
1 g |
Each for $94.05
|
|
||||||
AAH2911709
|
Thermo Scientific Chemicals
H2911709 |
10 g |
Each for $420.03
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5468-37-1 | |
171.62 | |
CVXGFPPAIUELDV-UHFFFAOYSA-N | |
2723597 | |
Cl.NCC(=O)C1=CC=CC=C1 |
C8H10ClNO | |
MFCD00012873 | |
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride | |
2-amino-1-phenylethanone;hydrochloride |
Specifications
5468-37-1 | |
Odorless | |
MFCD00012873 | |
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride | |
Cl.NCC(=O)C1=CC=CC=C1 | |
171.62 | |
171.63 | |
2-Aminoacetophenone hydrochloride |
∽194°C (decomposition) | |
C8H10ClNO | |
1 g | |
CVXGFPPAIUELDV-UHFFFAOYSA-N | |
2-amino-1-phenylethanone;hydrochloride | |
2723597 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 226-787-2
RTECSNumber : AM5940000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only