Learn More
2-(Aminomethyl)pyridine, 98+%, Thermo Scientific Chemicals
$56.62 - $357.93
Chemical Identifiers
CAS | 3731-51-9 |
---|---|
Molecular Formula | C6H8N2 |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00006360 |
InChI Key | WOXFMYVTSLAQMO-UHFFFAOYSA-N |
Synonym | 2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl |
PubChem CID | 19509 |
ChEBI | CHEBI:81387 |
SMILES | NCC1=CC=CC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2320806
|
Thermo Scientific Chemicals
B2320806 |
5 g |
Each for $56.62
|
|
|||||
AAB2320814
|
Thermo Scientific Chemicals
B2320814 |
25 g |
Each for $147.75
|
|
|||||
AAB2320822
|
Thermo Scientific Chemicals
B2320822 |
100 g |
Each for $357.93
|
|
|||||
Description
2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.
Solubility
Soluble in water (1000 mg/ml at 20°C), and ethanol.
Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, acids, air.
Chemical Identifiers
3731-51-9 | |
108.14 | |
WOXFMYVTSLAQMO-UHFFFAOYSA-N | |
19509 | |
NCC1=CC=CC=N1 |
C6H8N2 | |
MFCD00006360 | |
2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl | |
CHEBI:81387 |
Specifications
3731-51-9 | |
1.049 | |
202°C to 204°C | |
Amine-like | |
1.545 | |
5 g | |
108054 | |
2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl | |
WOXFMYVTSLAQMO-UHFFFAOYSA-N | |
pyridin-2-ylmethanamine | |
19509 | |
108.14 | |
2-(Aminomethyl)pyridine |
-20°C | |
11 to 12 | |
90°C (194°F) | |
C6H8N2 | |
MFCD00006360 | |
UN2735 | |
Air Sensitive | |
Soluble in water (1000mg/ml at 20°C),and ethanol. | |
NCC1=CC=CC=N1 | |
108.14 | |
CHEBI:81387 | |
99% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P374-P501c
H227-H302+H312-H314-H318-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 223-090-5
RTECSNumber : US1840000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only