Learn More
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 278030250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Aminoterephthalic acid | |
99.0 | |
324.0°C | |
Authentic | |
Glass bottle | |
MFCD00134536 | |
14, 558 | |
Solubility in water: slightly soluble. Other solubilities: insoluble in solvents | |
NC1=CC(=CC=C1C([O-])=O)C([O-])=O | |
179.13 | |
181.15 | |
Granular Powder |
10312-55-7 | |
100.0 | |
Yellow | |
98.5% min. (HPLC) | |
C8H5NO4 | |
25 g | |
aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751 | |
GPNNOCMCNFXRAO-UHFFFAOYSA-L | |
2-aminoterephthalic acid | |
2724822 | |
99% |
Chemical Identifiers
10312-55-7 | |
179.13 | |
GPNNOCMCNFXRAO-UHFFFAOYSA-L | |
2724822 | |
NC1=CC(=CC=C1C([O-])=O)C([O-])=O |
C8H5NO4 | |
MFCD00134536 | |
aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751 | |
2-aminoterephthalic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
RUO – Research Use Only