missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals

$141.84 - $446.75

Chemical Identifiers

CAS 10312-55-7
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00134536
InChI Key GPNNOCMCNFXRAO-UHFFFAOYSA-L
Synonym aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751
PubChem CID 2724822
IUPAC Name 2-aminoterephthalic acid
SMILES NC1=CC(=CC=C1C([O-])=O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC278030250
SDS View Documents
Thermo Scientific Chemicals
278030250
25 g Glass bottle
Each for $141.84
 
AC278031000
SDS View Documents
Thermo Scientific Chemicals
278031000
100 g Glass bottle
Each for $446.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10312-55-7
179.13
GPNNOCMCNFXRAO-UHFFFAOYSA-L
2724822
NC1=CC(=CC=C1C([O-])=O)C([O-])=O
C8H5NO4
MFCD00134536
aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751
2-aminoterephthalic acid

Specifications

10312-55-7
100.0
Yellow
98.5% min. (HPLC)
C8H5NO4
25 g
aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751
GPNNOCMCNFXRAO-UHFFFAOYSA-L
2-aminoterephthalic acid
2724822
99%
2-Aminoterephthalic acid
99.0
324.0°C
Authentic
Glass bottle
MFCD00134536
14, 558
Solubility in water: slightly soluble. Other solubilities: insoluble in solvents
NC1=CC(=CC=C1C([O-])=O)C([O-])=O
179.13
181.15
Granular Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only