missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Boc-amino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2902006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-(Boc-amino)benzeneboronic acid pinacol ester | |
98°C to 100°C | |
MFCD03411943 | |
tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-amino benzeneboronic acid pinacol ester, 2-n-boc-amino phenylboronic acid pinacol ester, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenylboronic acid pinacol ester, tert-butyl n-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NC(=O)OC(C)(C)C | |
319.208 | |
319.21 |
159624-15-4 | |
C17H26BNO4 | |
5 g | |
LVHGGVGVAUJQBB-UHFFFAOYSA-N | |
tert-butyl N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate | |
2773231 | |
97% |
Chemical Identifiers
159624-15-4 | |
319.208 | |
LVHGGVGVAUJQBB-UHFFFAOYSA-N | |
2773231 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NC(=O)OC(C)(C)C |
C17H26BNO4 | |
MFCD03411943 | |
tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-amino benzeneboronic acid pinacol ester, 2-n-boc-amino phenylboronic acid pinacol ester, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenylboronic acid pinacol ester, tert-butyl n-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester | |
tert-butyl N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only