missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Boc-amino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

$118.33 - $405.41

Chemical Identifiers

CAS 159624-15-4
Molecular Formula C17H26BNO4
Molecular Weight (g/mol) 319.208
MDL Number MFCD03411943
InChI Key LVHGGVGVAUJQBB-UHFFFAOYSA-N
Synonym tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-amino benzeneboronic acid pinacol ester, 2-n-boc-amino phenylboronic acid pinacol ester, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenylboronic acid pinacol ester, tert-butyl n-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester
PubChem CID 2773231
IUPAC Name tert-butyl N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2902003
View Documents
Thermo Scientific Chemicals
H2902003
1 g
Each for $118.33
 
AAH2902006
View Documents
Thermo Scientific Chemicals
H2902006
5 g
Each for $405.41
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

159624-15-4
319.208
LVHGGVGVAUJQBB-UHFFFAOYSA-N
2773231
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NC(=O)OC(C)(C)C
C17H26BNO4
MFCD03411943
tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-amino benzeneboronic acid pinacol ester, 2-n-boc-amino phenylboronic acid pinacol ester, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenylboronic acid pinacol ester, tert-butyl n-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester
tert-butyl N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Specifications

159624-15-4
C17H26BNO4
1 g
LVHGGVGVAUJQBB-UHFFFAOYSA-N
tert-butyl N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
2773231
97%
98°C to 100°C
MFCD03411943
tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-amino benzeneboronic acid pinacol ester, 2-n-boc-amino phenylboronic acid pinacol ester, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenylboronic acid pinacol ester, tert-butyl n-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NC(=O)OC(C)(C)C
319.208
319.21
2-(Boc-amino)benzeneboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only