missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Boc-amino)ethanol, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3235606

 View more versions of this product

Catalog No. AAH3235606


Explore available promotions

Description

Description

Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.

Solubility
Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-(Boc-amino)ethanol
92°C (0.22mmHg)
(CH3)3CCO2NHCH2CH2OH
MFCD00056657
Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol.
CC(C)(C)OC(=O)NCCO
tert-butyl N-(2-hydroxyethyl)carbamate
161.201
161.2
26690-80-2
C7H15NO3
1.449
n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
GPTXCAZYUMDUMN-UHFFFAOYSA-N
1.042
5 g
2733206
95%

Chemical Identifiers

26690-80-2
161.201
GPTXCAZYUMDUMN-UHFFFAOYSA-N
2733206
CC(C)(C)OC(=O)NCCO
C7H15NO3
MFCD00056657
n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
tert-butyl N-(2-hydroxyethyl)carbamate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only