Learn More
2-(Boc-amino)ethanol, 95%, Thermo Scientific Chemicals
$101.40 - $331.97
Chemical Identifiers
CAS | 26690-80-2 |
---|---|
Molecular Formula | C7H15NO3 |
Molecular Weight (g/mol) | 161.201 |
MDL Number | MFCD00056657 |
InChI Key | GPTXCAZYUMDUMN-UHFFFAOYSA-N |
Synonym | n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester |
PubChem CID | 2733206 |
IUPAC Name | tert-butyl N-(2-hydroxyethyl)carbamate |
SMILES | CC(C)(C)OC(=O)NCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3235606
|
Thermo Scientific Chemicals
H3235606 |
5 g |
Each for $101.40
|
|
|||||
AAH3235614
|
Thermo Scientific Chemicals
H3235614 |
25 g |
Each for $331.97
|
|
|||||
Description
Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSuzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.
Solubility
Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
26690-80-2 | |
161.201 | |
GPTXCAZYUMDUMN-UHFFFAOYSA-N | |
2733206 | |
CC(C)(C)OC(=O)NCCO |
C7H15NO3 | |
MFCD00056657 | |
n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester | |
tert-butyl N-(2-hydroxyethyl)carbamate |
Specifications
26690-80-2 | |
C7H15NO3 | |
MFCD00056657 | |
Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol. | |
GPTXCAZYUMDUMN-UHFFFAOYSA-N | |
tert-butyl N-(2-hydroxyethyl)carbamate | |
2733206 | |
95% | |
5 g |
92°C (0.22mmHg) | |
(CH3)3CCO2NHCH2CH2OH | |
n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester | |
1.449 | |
CC(C)(C)OC(=O)NCCO | |
161.201 | |
161.2 | |
1.042 | |
2-(Boc-amino)ethanol |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only