missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-(2-chloropyridin-4-yl)ethanone, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC53605DA

 View more versions of this product

Catalog No. CC53605DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-Bromo-1-(2-chloropyridin-4-yl)ethanone
94.5
95%
C7H5BrClNO
1g
BZWBRLWSWBQIBX-UHFFFAOYSA-N
2-bromo-1-(2-chloropyridin-4-yl)ethanone
16720431
95%
23794-16-3
100.0
Amber glass bottle
MFCD09702373
2-bromo-1-2-chloropyridin-4-yl ethanone, 2-bromo-1-2-chloro-4-pyridyl ethanone, 2-bromo-1-2-chloropyridin-4-yl ethan-1-one, 2-bromo-1-2-chloro-4-pyridinyl ethanone, 2-bromo-1-2-chloro-pyridin-4-yl-ethanone, 2-chloro-4-bromoacetylpyridine, 2-bromo-1-2-chloro-pyridin-4-yl ethanone, ethanone,2-bromo-1-2-chloro-4-pyridinyl, 2-bromo-1-2-chloro 4-pyridyl ethan-1-one, 2-bromanyl-1-2-chloranylpyridin-4-yl ethanone
C1=CN=C(C=C1C(=O)CBr)Cl
234.477
234.48

Chemical Identifiers

23794-16-3
234.477
BZWBRLWSWBQIBX-UHFFFAOYSA-N
16720431
C1=CN=C(C=C1C(=O)CBr)Cl
C7H5BrClNO
MFCD09702373
2-bromo-1-2-chloropyridin-4-yl ethanone, 2-bromo-1-2-chloro-4-pyridyl ethanone, 2-bromo-1-2-chloropyridin-4-yl ethan-1-one, 2-bromo-1-2-chloro-4-pyridinyl ethanone, 2-bromo-1-2-chloro-pyridin-4-yl-ethanone, 2-chloro-4-bromoacetylpyridine, 2-bromo-1-2-chloro-pyridin-4-yl ethanone, ethanone,2-bromo-1-2-chloro-4-pyridinyl, 2-bromo-1-2-chloro 4-pyridyl ethan-1-one, 2-bromanyl-1-2-chloranylpyridin-4-yl ethanone
2-bromo-1-(2-chloropyridin-4-yl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions