missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-(2-chloropyridin-4-yl)ethanone, 95%, Thermo Scientific™

Chemical Identifiers

CAS 23794-16-3
Molecular Formula C7H5BrClNO
Molecular Weight (g/mol) 234.477
MDL Number MFCD09702373
InChI Key BZWBRLWSWBQIBX-UHFFFAOYSA-N
Synonym 2-bromo-1-2-chloropyridin-4-yl ethanone, 2-bromo-1-2-chloro-4-pyridyl ethanone, 2-bromo-1-2-chloropyridin-4-yl ethan-1-one, 2-bromo-1-2-chloro-4-pyridinyl ethanone, 2-bromo-1-2-chloro-pyridin-4-yl-ethanone, 2-chloro-4-bromoacetylpyridine, 2-bromo-1-2-chloro-pyridin-4-yl ethanone, ethanone,2-bromo-1-2-chloro-4-pyridinyl, 2-bromo-1-2-chloro 4-pyridyl ethan-1-one, 2-bromanyl-1-2-chloranylpyridin-4-yl ethanone
PubChem CID 16720431
IUPAC Name 2-bromo-1-(2-chloropyridin-4-yl)ethanone
SMILES C1=CN=C(C=C1C(=O)CBr)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC53605CB
SDS View Documents
Thermo Scientific Chemicals
CC53605CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC53605DA
SDS View Documents
Thermo Scientific Chemicals
CC53605DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

23794-16-3
234.477
BZWBRLWSWBQIBX-UHFFFAOYSA-N
16720431
C1=CN=C(C=C1C(=O)CBr)Cl
C7H5BrClNO
MFCD09702373
2-bromo-1-2-chloropyridin-4-yl ethanone, 2-bromo-1-2-chloro-4-pyridyl ethanone, 2-bromo-1-2-chloropyridin-4-yl ethan-1-one, 2-bromo-1-2-chloro-4-pyridinyl ethanone, 2-bromo-1-2-chloro-pyridin-4-yl-ethanone, 2-chloro-4-bromoacetylpyridine, 2-bromo-1-2-chloro-pyridin-4-yl ethanone, ethanone,2-bromo-1-2-chloro-4-pyridinyl, 2-bromo-1-2-chloro 4-pyridyl ethan-1-one, 2-bromanyl-1-2-chloranylpyridin-4-yl ethanone
2-bromo-1-(2-chloropyridin-4-yl)ethanone

Specifications

23794-16-3
100.0
C7H5BrClNO
250mg
BZWBRLWSWBQIBX-UHFFFAOYSA-N
2-bromo-1-(2-chloropyridin-4-yl)ethanone
16720431
95%
94.5
95%
MFCD09702373
2-bromo-1-2-chloropyridin-4-yl ethanone, 2-bromo-1-2-chloro-4-pyridyl ethanone, 2-bromo-1-2-chloropyridin-4-yl ethan-1-one, 2-bromo-1-2-chloro-4-pyridinyl ethanone, 2-bromo-1-2-chloro-pyridin-4-yl-ethanone, 2-chloro-4-bromoacetylpyridine, 2-bromo-1-2-chloro-pyridin-4-yl ethanone, ethanone,2-bromo-1-2-chloro-4-pyridinyl, 2-bromo-1-2-chloro 4-pyridyl ethan-1-one, 2-bromanyl-1-2-chloranylpyridin-4-yl ethanone
C1=CN=C(C=C1C(=O)CBr)Cl
234.477
234.48
2-Bromo-1-(2-chloropyridin-4-yl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'